Assay Details
| Gene Symbol: GRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258059.1 | NP_001244988.1 | 279 | 139 | ||
| XM_028835529.1 | XP_028691362.1 | 409 | 139 | |||
| GenBank mRNA | JU474060.1 | - | 279 | 139 | ||
More Information
Set Membership: |
|

GRAP
Genomic Map