Assay Details
| Gene Symbol: TXNL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260866.1 | NP_001247795.1 | 203 | 151 | ||
| GenBank mRNA | JU319794.1 | - | 272 | 151 | ||
| JU473637.1 | - | 297 | 151 | |||
| JV046000.1 | - | 203 | 151 | |||
| JV529929.1 | - | 259 | 151 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TXNL4A
Genomic Map