Assay Details
| Gene Symbol: RAB32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193894.3 | NP_001180823.1 | 421 | 92 | ||
| GenBank mRNA | JU332665.1 | - | 272 | 92 | ||
| JV640841.1 | - | 307 | 92 | |||
More Information
Set Membership: |
|

RAB32
Genomic Map