Assay Details
| Gene Symbol: PLAAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257427.1 | NP_001244356.1 | 794 | 105 | ||
| XM_015113770.2 | XP_014969256.1 | 631 | 105 | |||
| GenBank mRNA | JU332716.1 | - | 794 | 105 | ||
| JU473148.1 | - | 450 | 105 | |||
| JV043551.1 | - | 537 | 105 | |||
| JV604480.1 | - | 538 | 105 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLAAT3
Genomic Map