Assay Details
| Gene Symbol: UGT1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032869.1 | NP_001028041.1 | 1003 | 147 | ||
| XM_015111222.2 | XP_014966708.2 | 1037 | 147 | |||
| XM_015111223.2 | XP_014966709.2 | 1156 | 147 | |||
| GenBank mRNA | AF360121.1 | - | 1003 | 147 | ||
| AY029169.1 | - | 1003 | 147 | |||
| JU329361.1 | - | 147 | ||||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UGT1A1
Genomic Map