Assay Details
| Gene Symbol: DCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265650.1 | NP_001252579.1 | 213 | 83 | ||
| XM_015117359.2 | XP_014972845.1 | 243 | 83 | |||
| XM_015117360.2 | XP_014972846.1 | 243 | 83 | |||
| GenBank mRNA | JU326234.1 | - | 89 | 83 | ||
| JV667335.1 | - | 213 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DCTN3
Genomic Map