Assay Details
| Gene Symbol: KLF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842551.1 | XP_028698384.1 | 1215 | 96 | ||
| XM_028842552.1 | XP_028698385.1 | 1275 | 96 | |||
| XM_028842553.1 | XP_028698386.1 | 823 | 96 | |||
| GenBank mRNA | JV635739.1 | - | 1501 | 96 | ||
More Information
Set Membership: |
|

KLF8
Genomic Map