Assay Details
| Gene Symbol: POU6F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133684.2 | XP_014989170.2 | 1542 | 50 | ||
| XM_028845856.1 | XP_028701689.1 | 1399 | 50 | |||
More Information
Set Membership: |
|

POU6F2
Genomic Map