Assay Details
| Gene Symbol: CD300A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090848.4 | XP_001090848.3 | 999 | 104 | ||
| XM_015120268.2 | XP_014975754.2 | 999 | 104 | |||
| GenBank mRNA | JU479591.1 | - | 649 | 104 | ||
More Information
Set Membership: |
|

CD300A
Genomic Map