Assay Details
| Gene Symbol: CA14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257903.2 | NP_001244832.1 | 362 | 84 | ||
| XM_028843154.1 | XP_028698987.1 | 1209 | 84 | |||
| XM_028843156.1 | XP_028698989.1 | 1209 | 84 | |||
| GenBank mRNA | JU323034.1 | - | 538 | 84 | ||
| JU323035.1 | - | 366 | 84 | |||
More Information
Set Membership: |
|

CA14
Genomic Map