Assay Details
| Gene Symbol: FBXO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096104.3 | XP_001096104.1 | 1067 | 94 | ||
| XM_015137103.2 | XP_014992589.1 | 1394 | 94 | |||
| GenBank mRNA | JU478171.1 | - | 1041 | 94 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FBXO5
Genomic Map