Assay Details
| Gene Symbol: NUBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266874.2 | NP_001253803.1 | 176 | 93 | ||
| GenBank mRNA | JU332287.1 | - | 176 | 93 | ||
| JV635895.1 | - | 172 | 93 | |||
More Information
Set Membership: |
|

NUBP2
Genomic Map