Assay Details
| Gene Symbol: LAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194698.2 | NP_001181627.2 | 464 | 90 | ||
| XM_028827442.1 | XP_028683275.1 | 911 | 90 | |||
| GenBank mRNA | JU333761.1 | - | 362 | 90 | ||
| JV567601.1 | - | 465 | 90 | |||
| JV601809.1 | - | 461 | 90 | |||
| JV678913.1 | - | 342 | 90 | |||
More Information
Set Membership: |
|

LAMP5
Genomic Map