Assay Details
| Gene Symbol: QPCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101173.4 | XP_001101173.3 | 905 | 62 | ||
| XM_015112121.2 | XP_014967607.2 | 1054 | 62 | |||
| XM_028831756.1 | XP_028687589.1 | 989 | 62 | |||
More Information
Set Membership: |
|

QPCT
Genomic Map