Assay Details
| Gene Symbol: DNAJC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001092902.4 | XP_001092902.2 | 589 | 80 | ||
| GenBank mRNA | JU334164.1 | - | 122 | 80 | ||
| JU475289.1 | - | 141 | 80 | |||
| JV614408.1 | - | 128 | 80 | |||
More Information
Set Membership: |
|

DNAJC15
Genomic Map