Assay Details
| Gene Symbol: PYCARD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194572.1 | NP_001181501.1 | 315 | 147 | ||
| GenBank mRNA | JU334167.1 | - | 293 | 147 | ||
| JU475292.1 | - | 379 | 147 | |||
| JV045207.1 | - | 302 | 147 | |||
| JV649375.1 | - | 401 | 147 | |||
More Information
Set Membership: |
|

PYCARD
Genomic Map