Assay Details
| Gene Symbol: TIMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835843.1 | XP_028691676.1 | 1121 | 122 | ||
| GenBank mRNA | JU325556.1 | - | 433 | 122 | ||
| JU475336.1 | - | 447 | 122 | |||
| JV046885.1 | - | 447 | 122 | |||
| JV566111.1 | - | 454 | 122 | |||
| JV601037.1 | - | 463 | 122 | |||
More Information
Set Membership: |
|

TIMM22
Genomic Map