Assay Details
| Gene Symbol: NDUFA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001089493.4 | XP_001089493.1 | 526 | 120 | ||
| GenBank mRNA | JU323227.1 | - | 485 | 120 | ||
| JV045225.1 | - | 518 | 120 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NDUFA8
Genomic Map