Assay Details
| Gene Symbol: IL1RAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127123.2 | XP_014982609.1 | 1729 | 94 | ||
| GenBank mRNA | JU320661.1 | - | 1205 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL1RAPL1| Gene Symbol: IL1RAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127123.2 | XP_014982609.1 | 1729 | 94 | ||
| GenBank mRNA | JU320661.1 | - | 1205 | 94 | ||
Set Membership: |
|