Assay Details
| Gene Symbol: FAM162A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261743.2 | NP_001248672.1 | 336 | 117 | ||
| XM_015129915.2 | XP_014985401.1 | 476 | 117 | |||
| XM_015129916.2 | XP_014985402.1 | 371 | 117 | |||
| GenBank mRNA | JU333956.1 | - | 359 | 117 | ||
| JU475144.1 | - | 329 | 117 | |||
| JV046734.1 | - | 338 | 117 | |||
| JV464514.1 | - | 256 | 117 | |||
| JV612980.1 | - | 361 | 117 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FAM162A
Genomic Map