Assay Details
| Gene Symbol: RHOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113598.2 | XP_014969084.2 | 560 | 114 | ||
| XM_015113599.2 | XP_014969085.2 | 362 | 114 | |||
| GenBank mRNA | JV635808.1 | - | 484 | 114 | ||
More Information
Set Membership: |
|

RHOD
Genomic Map