Assay Details
Gene Symbol: GARRE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193886.2 | NP_001180815.1 | 1795 | 105 | ||
XM_015123517.2 | XP_014979003.1 | 3351 | 105 | |||
XM_015123518.2 | XP_014979004.1 | 1982 | 105 | |||
XM_028838424.1 | XP_028694257.1 | 1942 | 105 | |||
XM_028838426.1 | XP_028694259.1 | 1364 | 105 | |||
GenBank mRNA | JU321568.1 | - | 1784 | 105 | ||
JU470676.1 | - | 1781 | 105 | |||
JV045078.1 | - | 1675 | 105 | |||
JV636941.1 | - | 1777 | 105 |

More Information
Set Membership: |
![]() ![]() |
