Assay Details
| Gene Symbol: DHRS7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261025.1 | NP_001247954.1 | 797 | 95 | ||
| XM_015118687.2 | XP_014974173.2 | 770 | 95 | |||
| XM_015118688.2 | XP_014974174.2 | 1088 | 95 | |||
| XM_028835541.1 | XP_028691374.1 | 952 | 95 | |||
| XM_028835542.1 | XP_028691375.1 | 1215 | 95 | |||
| XM_028835543.1 | XP_028691376.1 | 1173 | 95 | |||
| GenBank mRNA | JU322892.1 | - | 797 | 95 | ||
More Information
Set Membership: |
|

DHRS7B
Genomic Map