Assay Details
Gene Symbol: NOC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143132.2 | XP_014998618.2 | 1912 | 71 | ||
XM_015143643.2 | XP_014999129.2 | 1912 | 71 | |||
GenBank mRNA | JV046723.1 | - | 1872 | 71 | ||
JV612825.1 | - | 1861 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
