Assay Details
| Gene Symbol: DESI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266945.1 | NP_001253874.1 | 479 | 111 | ||
| XM_015150401.2 | XP_015005887.1 | 592 | 111 | |||
| GenBank mRNA | JU320969.1 | - | 479 | 111 | ||
| JU472380.1 | - | 477 | 111 | |||
| JV044554.1 | - | 479 | 111 | |||
More Information
Set Membership: |
|

DESI1
Genomic Map