Assay Details
| Gene Symbol: CRLF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257619.1 | NP_001244548.1 | 1149 | 137 | ||
| GenBank mRNA | JU334874.1 | - | 1149 | 137 | ||
| JU475893.1 | - | 1172 | 137 | |||
| JV045732.1 | - | 1177 | 137 | |||
| JV653292.1 | - | 1154 | 137 | |||
More Information
Set Membership: |
|

CRLF3
Genomic Map