Assay Details
| Gene Symbol: METTL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125785.2 | XP_014981271.1 | 611 | 127 | ||
| XM_015125786.2 | XP_014981272.1 | 611 | 127 | |||
| XM_015125788.2 | XP_014981274.1 | 611 | 127 | |||
| XM_028841230.1 | XP_028697063.1 | 611 | 127 | |||
| GenBank mRNA | JU334785.1 | - | 416 | 127 | ||
| JU471813.1 | - | 550 | 127 | |||
| JV047250.1 | - | 427 | 127 | |||
| JV565039.1 | - | 518 | 127 | |||
| JV600708.1 | - | 539 | 127 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

METTL9
Genomic Map