Assay Details
| Gene Symbol: LACTB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145553.1 | XP_015001039.1 | 362 | 91 | ||
| GenBank mRNA | JU322747.1 | - | 202 | 91 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LACTB2
Genomic Map