Assay Details
| Gene Symbol: DESI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257794.1 | NP_001244723.1 | 458 | 116 | ||
| GenBank mRNA | JU334765.1 | - | 458 | 116 | ||
| JU475806.1 | - | 497 | 116 | |||
| JV652911.1 | - | 504 | 116 | |||
More Information
Set Membership: |
|

DESI2
Genomic Map