Assay Details
| Gene Symbol: TIMMDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194452.2 | NP_001181381.1 | 873 | 155 | ||
| GenBank mRNA | JU323495.1 | - | 884 | 155 | ||
| JU487800.1 | - | 912 | 155 | |||
| JV045055.1 | - | 864 | 155 | |||
| JV528414.1 | - | 897 | 155 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TIMMDC1
Genomic Map