Assay Details
| Gene Symbol: COMMD8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260885.1 | NP_001247814.1 | 551 | 132 | ||
| XM_015138318.2 | XP_014993804.1 | 602 | 132 | |||
| XM_015138319.2 | XP_014993805.1 | 1022 | 132 | |||
| GenBank mRNA | JU476157.1 | - | 551 | 132 | ||
| JV717760.1 | - | 500 | 132 | |||
More Information
Set Membership: |
|

COMMD8
Genomic Map