Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266028.1 | NP_001252957.1 | 984 | 70 | ||
| XM_015128700.2 | XP_014984186.1 | 1205 | 70 | |||
| GenBank mRNA | JV064637.1 | - | 623 | 70 | ||
| JV635844.1 | - | 984 | 70 | |||
More Information
Set Membership: |
|

TMEM45A
Genomic Map