Assay Details
| Gene Symbol: ETNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260518.1 | NP_001247447.1 | 906 | 135 | ||
| XM_015118445.2 | XP_014973931.1 | 1110 | 135 | |||
| XM_015118455.2 | XP_014973941.1 | 987 | 135 | |||
| XM_015118477.2 | XP_014973963.1 | 850 | 135 | |||
| XM_028835182.1 | XP_028691015.1 | 1110 | 135 | |||
| XM_028835242.1 | XP_028691075.1 | 987 | 135 | |||
| XM_028835302.1 | XP_028691135.1 | 1110 | 135 | |||
| XM_028835329.1 | XP_028691162.1 | 1110 | 135 | |||
| GenBank mRNA | JU325863.1 | - | 597 | 135 | ||
| JV047529.1 | - | 906 | 135 | |||
| JV655197.1 | - | 1077 | 135 | |||
More Information
Set Membership: |
|

ETNK2
Genomic Map