Assay Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261114.1 | NP_001248043.1 | 1298 | 107 | ||
| GenBank mRNA | JU322717.1 | - | 1146 | 107 | ||
| JU470866.1 | - | 1419 | 107 | |||
| JV045191.1 | - | 1298 | 107 | |||
| JV637271.1 | - | 1438 | 107 | |||
More Information
Set Membership: |
|

TMEM19
Genomic Map