Assay Details
| Gene Symbol: SYNJ2BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260960.2 | NP_001247889.1 | 154 | 117 | ||
| GenBank mRNA | JU322556.1 | - | 171 | 117 | ||
| JU470553.1 | - | 144 | 117 | |||
| JV044006.1 | - | 158 | 117 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260960.2 transcripts.
Set Membership: |
|

SYNJ2BP
Genomic Map