Assay Details
| Gene Symbol: SLC39A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194438.3 | NP_001181367.1 | 1639 | 138 | ||
| XM_001109283.4 | XP_001109283.1 | 1370 | 138 | |||
| XM_015143935.2 | XP_014999421.2 | 1031 | 138 | |||
| XM_015143937.2 | XP_014999423.1 | 692 | 138 | |||
| GenBank mRNA | JU321306.1 | - | 1306 | 138 | ||
| JU471099.1 | - | 1346 | 138 | |||
| JV655379.1 | - | 1360 | 138 | |||
More Information
Set Membership: |
|

SLC39A9
Genomic Map