Assay Details
| Gene Symbol: ZGRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139049.2 | XP_014994535.2 | 2796 | 93 | ||
| XM_015139050.2 | XP_014994536.2 | 2738 | 93 | |||
| XM_015139051.2 | XP_014994537.2 | 2796 | 93 | |||
| XM_015139053.2 | XP_014994539.2 | 2796 | 93 | |||
| XM_015139054.2 | XP_014994540.2 | 2796 | 93 | |||
| XM_015139055.2 | XP_014994541.2 | 2796 | 93 | |||
More Information
Set Membership: |
|

ZGRF1
Genomic Map