Assay Details
| Gene Symbol: GABRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093973.3 | XP_001093973.1 | 833 | 86 | ||
| GenBank mRNA | JU325846.1 | - | 527 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GABRQ| Gene Symbol: GABRQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093973.3 | XP_001093973.1 | 833 | 86 | ||
| GenBank mRNA | JU325846.1 | - | 527 | 86 | ||
Set Membership: |
|