Assay Details
| Gene Symbol: CYTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194003.2 | NP_001180932.2 | 150 | 72 | ||
| XM_028848102.1 | XP_028703935.1 | 252 | 72 | |||
| GenBank mRNA | JU476067.1 | - | 175 | 72 | ||
More Information
Set Membership: |
|

CYTL1
Genomic Map