Assay Details
| Gene Symbol: MYDGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122545.2 | XP_014978031.1 | 468 | 129 | ||
| GenBank mRNA | JU322163.1 | - | 401 | 129 | ||
| JU470815.1 | - | 379 | 129 | |||
| JV622307.1 | - | 347 | 129 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MYDGF
Genomic Map