Assay Details
| Gene Symbol: KMT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028830088.1 | XP_028685921.1 | 715 | 122 | ||
| XM_028830089.1 | XP_028685922.1 | 715 | 122 | |||
| XM_028830090.1 | XP_028685923.1 | 211 | 122 | |||
| XM_028830091.1 | XP_028685924.1 | 593 | 122 | |||
| XM_028830092.1 | XP_028685925.1 | 418 | 122 | |||
| GenBank mRNA | JU326980.1 | - | 232 | 122 | ||
| JU326981.1 | - | 232 | 122 | |||
| JU326982.1 | - | 232 | 122 | |||
| JU486277.1 | - | 285 | 122 | |||
| JU486278.1 | - | 285 | 122 | |||
More Information
Set Membership: |
|

KMT5A
Genomic Map