Assay Details
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257750.1 | NP_001244679.1 | 430 | 67 | ||
GenBank mRNA | JU334669.1 | - | 430 | 67 | ||
JU475718.1 | - | 439 | 67 | |||
JV047163.1 | - | 417 | 67 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
