Assay Details
| Gene Symbol: TSKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115406.4 | XP_001115406.2 | 658 | 73 | ||
| XM_015124607.2 | XP_014980093.2 | 659 | 73 | |||
| XM_015124608.2 | XP_014980094.2 | 658 | 73 | |||
| XM_015124610.2 | XP_014980096.2 | 658 | 73 | |||
| XM_028838780.1 | XP_028694613.1 | 658 | 73 | |||
| XM_028838781.1 | XP_028694614.1 | 658 | 73 | |||
More Information
Set Membership: |
|

TSKS
Genomic Map