Assay Details
| Gene Symbol: C16H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257907.1 | NP_001244836.1 | 694 | 136 | ||
| GenBank mRNA | JU322894.1 | - | 694 | 136 | ||
| JU473688.1 | - | 685 | 136 | |||
| JV044191.1 | - | 693 | 136 | |||
| JV590536.1 | - | 319 | 136 | |||
More Information
Set Membership: |
|

C16H17orf75
Genomic Map