Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265990.1 | NP_001252919.1 | 1175 | 144 | ||
| XM_015140671.2 | XP_014996157.1 | 1143 | 144 | |||
| XM_015140672.2 | XP_014996158.1 | 1486 | 144 | |||
| XM_028849172.1 | XP_028705005.1 | 1146 | 144 | |||
| XM_028849173.1 | XP_028705006.1 | 1489 | 144 | |||
| XM_028849174.1 | XP_028705007.1 | 1446 | 144 | |||
| XM_028849175.1 | XP_028705008.1 | 1179 | 144 | |||
| XM_028849177.1 | XP_028705010.1 | 2185 | 144 | |||
| GenBank mRNA | JU321037.1 | - | 1175 | 144 | ||
| JU322547.1 | - | 1218 | 144 | |||
More Information
Set Membership: |
|

GRAMD2B
Genomic Map