Assay Details
| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123230.2 | XP_014978716.1 | 560 | 73 | ||
| XM_028840033.1 | XP_028695866.1 | 404 | 73 | |||
| GenBank mRNA | JU336340.1 | - | 480 | 73 | ||
| JU472740.1 | - | 534 | 73 | |||
| JV045308.1 | - | 537 | 73 | |||
| JV601144.1 | - | 540 | 73 | |||
| JV668200.1 | - | 538 | 73 | |||
More Information
Set Membership: |
|

TMEM38A
Genomic Map