Assay Details
| Gene Symbol: UBL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261050.1 | NP_001247979.1 | 258 | 99 | ||
| XM_015122785.2 | XP_014978271.1 | 296 | 99 | |||
| XM_028838446.1 | XP_028694279.1 | 423 | 99 | |||
| GenBank mRNA | JU471006.1 | - | 258 | 99 | ||
| JV601589.1 | - | 232 | 99 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

UBL5
Genomic Map