Assay Details
| Gene Symbol: NDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193470.2 | NP_001180399.2 | 312 | 108 | ||
| XM_015138959.2 | XP_014994445.2 | 858 | 108 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDNF| Gene Symbol: NDNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193470.2 | NP_001180399.2 | 312 | 108 | ||
| XM_015138959.2 | XP_014994445.2 | 858 | 108 | |||
Set Membership: |
|