Assay Details
| Gene Symbol: FN3KRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194815.3 | NP_001181744.2 | 169 | 121 | ||
| GenBank mRNA | JV048260.1 | - | 169 | 121 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FN3KRP
Genomic Map