Assay Details
| Gene Symbol: MANEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136962.2 | XP_014992448.1 | 726 | 100 | ||
| XM_028847340.1 | XP_028703173.1 | 299 | 100 | |||
| GenBank mRNA | JU322720.1 | - | 679 | 100 | ||
| JU470949.1 | - | 686 | 100 | |||
| JV704005.1 | - | 691 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028847340.1 transcripts.
Set Membership: |
|

MANEA
Genomic Map